siRNA Details
Gene Symbol: Pla2g5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017174.1 | NP_058870.1 | 4 | 448 | ||
XM_006239139.2 | XP_006239201.1 | 5 | 1247 | |||
XM_006239140.2 | XP_006239202.1 | 5 | 1826 | |||
XM_006239141.2 | XP_006239203.1 | 4 | 370 | |||
XM_008764209.1 | XP_008762431.1 | 6 | 1361 | |||
XM_008764210.1 | XP_008762432.1 | 4 | 2468 | |||
XM_008764211.1 | XP_008762433.1 | 3 | 483 | |||
GenBank mRNA | AY651028.1 | - | 4 | 315 | ||
BC085745.1 | - | 4 | 473 | 7192819 | ||
U03763.1 | - | 4 | 448 |
