siRNA Details
Gene Symbol: Ptk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017318.2 | NP_059014.2 | 7 | 739 | ||
XM_006252143.2 | XP_006252205.1 | 7 | 1064 | |||
XM_006252144.2 | XP_006252206.1 | 7 | 4055 | |||
XM_006252145.2 | XP_006252207.1 | 7 | 868 | |||
XM_008770773.1 | XP_008768995.1 | 7 | 2252 | |||
XM_008770774.1 | XP_008768996.1 | 7 | 1818 | |||
GenBank mRNA | BC101921.1 | - | 7 | 739 | 7937929 | |
D45854.1 | - | 7 | 917 | |||
U69109.1 | - | 6 | 659 |
