siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 4 | 612 | ||
XM_006250069.2 | XP_006250131.1 | 6 | 1929 | |||
GenBank mRNA | AY029412.1 | - | 4 | 612 | ||
L15453.1 | - | 4 | 652 |
