siRNA Details
Gene Symbol: Eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020075.1 | NP_064460.1 | 3 | 422 | ||
XM_006240608.1 | XP_006240670.1 | 5 | 685 | |||
XM_006240609.2 | XP_006240671.1 | 4 | 965 | |||
GenBank mRNA | BC062398.1 | - | 4 | 610 | 5623373 | |
FQ222051.1 | - | 1 | 502 | |||
L11651.1 | - | 3 | 422 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173363, NM_178041, AK051980, BC019769, BC025523, and others.
Gene Ontology Categories:
