siRNA Details
Gene Symbol: Notch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020087.2 | NP_064472.2 | 30 | 5563 | ||
GenBank mRNA | AF164486.2 | - | 30 | 5563 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Notch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020087.2 | NP_064472.2 | 30 | 5563 | ||
GenBank mRNA | AF164486.2 | - | 30 | 5563 |