siRNA Details
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021663.2 | NP_067695.1 | 4 | 443 | ||
XM_006230095.2 | XP_006230157.1 | 3 | 351 | |||
GenBank mRNA | AF238223.1 | - | 2 | 230 | ||
AF250142.1 | - | 3 | 330 | |||
BC061778.1 | - | 4 | 443 | 5621784 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001130479, AJ222586, AK145934, AK160442, BC010459.
Gene Ontology Categories:
