siRNA Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021762.1 | NP_068530.1 | 3 | 295 | ||
XM_008769496.1 | XP_008767718.1 | 3 | 314 | |||
GenBank mRNA | AF262356.1 | - | 3 | 295 | ||
AY325258.1 | - | 370 |
