siRNA Details
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007019.3 | NP_008950.1 | 4 | 361 | ||
NM_181800.2 | NP_861516.1 | 3 | 274 | |||
NM_181801.3 | NP_861517.1 | 4 | 398 | |||
GenBank mRNA | AK311949.1 | - | 4 | 328 | ||
BC007656.2 | - | 4 | 284 | 3345575 | ||
BC016292.1 | - | 4 | 290 | 3910286 | ||
BC028161.1 | - | 3 | 900 | |||
BC050736.1 | - | 4 | 311 | 6503915 | ||
BF974975.1 | - | 417 | ||||
BI858659.1 | - | 4 | 398 | |||
BM747513.1 | - | 4 | 316 | |||
BP227692.1 | - | 4 | 388 | |||
BT007300.1 | - | 4 | 241 | |||
BT020165.1 | - | 4 | 241 | |||
BT020166.1 | - | 4 | 241 | |||
BU844974.1 | - | 3 | 176 | |||
CK904708.1 | - | 3 | 186 | |||
CR456988.1 | - | 4 | 241 | |||
DB062573.1 | - | 4 | 362 | |||
U73379.1 | - | 4 | 281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
