siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 2, 3 | 250 | ||
XM_008760546.1 | XP_008758768.1 | 3, 4 | 843 | |||
XM_008760547.1 | XP_008758769.1 | 2, 3 | 1225 | |||
XM_008760548.1 | XP_008758770.1 | 2, 3 | 552 | |||
XM_008760549.1 | XP_008758771.1 | 3, 4 | 432 | |||
XM_008760550.1 | XP_008758772.1 | 2, 3 | 360 | |||
GenBank mRNA | BC070936.1 | - | 2, 3 | 251 | 7099111 |
