siRNA Details
Gene Symbol: Vps33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022286.2 | NP_071622.1 | 19, 20 | 1782 | ||
XM_008759557.1 | XP_008757779.1 | 16, 17 | 1219 | |||
GenBank mRNA | BC081707.1 | - | 19, 20 | 1782 | 7128590 | |
U35245.1 | - | 19, 20 | 1497 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178070, BC034170, BC028783, AK032076.
Gene Ontology Categories:
