siRNA Details
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022298.1 | NP_071634.1 | 2, 3 | 288 | ||
GenBank mRNA | BC062238.1 | - | 2, 3 | 322 | 6922126 | |
BC078830.1 | - | 2, 3 | 301 | 7115494 | ||
FQ210110.1 | - | 2, 3 | 320 | |||
FQ212756.1 | - | 2, 3 | 254 | |||
FQ213186.1 | - | 2, 3 | 320 | |||
V01227.1 | - | 2, 3 | 288 |
Gene Symbol: LOC100909441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749354.3 | XP_003749402.1 | 2, 3 | 315 |
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044270.2 | NP_001037735.1 | 2, 3 | 321 | ||
GenBank mRNA | BC060572.1 | - | 2, 3 | 325 | 6920989 | |
BC076379.1 | - | 2, 3 | 307 | 7106502 | ||
FQ212426.1 | - | 2, 3 | 316 | |||
FQ212740.1 | - | 2, 3 | 316 | |||
FQ213098.1 | - | 2, 3 | 321 | |||
FQ213226.1 | - | 2, 3 | 316 | |||
FQ213439.1 | - | 2, 3 | 316 | |||
FQ213947.1 | - | 2, 3 | 316 | |||
FQ214075.1 | - | 2, 3 | 319 | |||
FQ214100.1 | - | 2, 3 | 316 | |||
FQ214301.1 | - | 2, 3 | 319 | |||
FQ223081.1 | - | 2, 3 | 319 | |||
FQ225046.1 | - | 2, 3 | 321 | |||
FQ225339.1 | - | 2, 3 | 316 | |||
FQ225425.1 | - | 2, 3 | 316 | |||
FQ225650.1 | - | 2, 3 | 316 | |||
FQ228903.1 | - | 103 | ||||
FQ232160.1 | - | 1, 2 | 226 | |||
FQ232527.1 | - | 2, 3 | 320 | |||
FQ233000.1 | - | 2, 3 | 318 | |||
FQ233566.1 | - | 2, 3 | 319 | |||
FQ234135.1 | - | 2, 3 | 316 |
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011995.1 | NP_001011995.1 | 2, 3 | 304 | ||
GenBank mRNA | BC078829.1 | - | 2, 3 | 304 | 7112420 | |
FQ221879.1 | - | 1, 2 | 94 | |||
FQ225183.1 | - | 2, 3 | 325 | |||
FQ225366.1 | - | 2, 3 | 324 | |||
FQ226218.1 | - | 2, 3 | 325 | |||
FQ235315.1 | - | 2, 3 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
