siRNA Details
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022387.3 | NP_071782.1 | 5 | 479 | ||
XM_006242957.2 | XP_006243019.1 | 5 | 945 | |||
XM_006242958.2 | XP_006243020.1 | 5 | 478 | |||
XM_008766172.1 | XP_008764394.1 | 5 | 709 | |||
GenBank mRNA | AF016048.1 | - | 5 | 442 | ||
BC081714.1 | - | 5 | 479 | 7108923 | ||
FQ212439.1 | - | 5 | 766 |
