siRNA Details
Gene Symbol: Qdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022390.1 | NP_071785.1 | 7 | 809 | ||
GenBank mRNA | BC072536.1 | - | 7 | 822 | 7103182 | |
FQ209462.1 | - | 5 | 552 | |||
FQ213296.1 | - | 7 | 788 | |||
FQ213843.1 | - | 7 | 838 | |||
J03481.1 | - | 7 | 809 |
