siRNA Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033898.1 | NP_001029070.1 | 2 | 130 | ||
NM_022535.2 | NP_071980.2 | 2 | 130 | |||
GenBank mRNA | AJ001929.1 | - | 2 | 163 | ||
BC101908.1 | - | 2 | 130 | 7933832 | ||
CK476875.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
