siRNA Details
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022689.2 | NP_073180.1 | 4 | 268 | ||
GenBank mRNA | AF052596.1 | - | 120 | |||
BC127494.1 | - | 4 | 268 | 8361815 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022689.2 | NP_073180.1 | 4 | 268 | ||
GenBank mRNA | AF052596.1 | - | 120 | |||
BC127494.1 | - | 4 | 268 | 8361815 |