siRNA Details
Gene Symbol: Exoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022691.1 | NP_073182.1 | 2, 3 | 416 | ||
GenBank mRNA | AF032667.1 | - | 2, 3 | 416 | ||
FQ231273.1 | - | 183 |
Gene Symbol: LOC100911734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768426.1 | XP_008766648.1 | 2, 3 | 197 | ||
XM_008768427.1 | XP_008766649.1 | 2, 3 | 197 | |||
XM_008768428.1 | XP_008766650.1 | 2, 3 | 197 | |||
XM_008768429.1 | XP_008766651.1 | 2, 3 | 197 | |||
XM_008768430.1 | XP_008766652.1 | 197 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF014461.
Gene Ontology Categories:
