siRNA Details
Gene Symbol: Mgat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023095.1 | NP_075583.1 | 3 | 466 | ||
GenBank mRNA | L14284.1 | - | 3 | 466 |
Gene Symbol: LOC100909582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249676.2 | XP_006249738.1 | 3 | 818 | ||
XM_006249677.2 | XP_006249739.1 | 3 | 815 | |||
XM_006249679.2 | XP_006249741.1 | 2 | 390 | |||
XM_008769509.1 | XP_008767731.1 | 4 | 966 |
