siRNA Details
Gene Symbol: CALM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006888.4 | NP_008819.1 | 6 | 1050 | ||
XM_006720258.3 | XP_006720321.1 | 6 | 1679 | |||
GenBank mRNA | BC000454.2 | - | 6 | 984 | 2821489 | |
BC007965.2 | - | 6 | 1024 | 4308380 | ||
BC008597.1 | - | 6 | 984 | 3452971 | ||
BC011834.2 | - | 6 | 1004 | 3942245 | ||
BX537677.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
