siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024146.1 | NP_077060.1 | 2 | 235 | ||
XM_006253323.2 | XP_006253385.1 | 2 | 243 | |||
XM_006253324.2 | XP_006253386.1 | 3 | 456 | |||
GenBank mRNA | D12498.1 | - | 2 | 235 |
