siRNA Details
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024371.2 | NP_077347.1 | 2 | 235 | ||
XM_006237059.1 | XP_006237121.1 | 3 | 444 | |||
XM_006237060.2 | XP_006237122.1 | 3 | 2664 | |||
GenBank mRNA | M59742.1 | - | 2 | 235 |
