siRNA Details
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282279.1 | NP_001269208.1 | 7 | 1733 | ||
NM_001282280.1 | NP_001269209.1 | 8 | 1826 | |||
NM_001282281.1 | NP_001269210.1 | 8 | 1819 | |||
NM_006907.3 | NP_008838.2 | 7 | 2155 | |||
NM_153824.2 | NP_722546.1 | 8 | 1812 | |||
XM_005256381.2 | XP_005256438.1 | 8 | 1784 | |||
XM_011523583.2 | XP_011521885.1 | 8 | 2023 | |||
XM_011523584.2 | XP_011521886.1 | 8 | 1778 | |||
XM_011523585.2 | XP_011521887.1 | 7 | 1649 | |||
XM_017024909.1 | XP_016880398.1 | 7 | 1662 | |||
GenBank mRNA | AF218000.1 | - | 8 | 1648 | ||
AK225879.1 | - | 7 | 1701 | |||
BC001504.2 | - | 7 | 1752 | 3505512 | ||
BC022244.1 | - | 7 | 1673 | 4420238 | ||
BC071842.1 | - | 7 | 1774 | 5088067 | ||
M77836.1 | - | 7 | 1723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
