siRNA Details
Gene Symbol: Avil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024401.2 | NP_077377.2 | 7, 8 | 867 | ||
XM_008765423.1 | XP_008763645.1 | 6, 7 | 999 | |||
GenBank mRNA | AF099929.1 | - | 7, 8 | 874 |
