siRNA Details
Gene Symbol: Acvr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024486.1 | NP_077812.1 | 3 | 242 | ||
XM_006234214.2 | XP_006234276.1 | 5 | 1847 | |||
XM_008761862.1 | XP_008760084.1 | 6 | 1904 | |||
XM_008761863.1 | XP_008760085.1 | 6 | 4148 | |||
XM_008761864.1 | XP_008760086.1 | 6 | 3894 | |||
XM_008761865.1 | XP_008760087.1 | 6 | 3937 | |||
XM_008761866.1 | XP_008760088.1 | 6 | 614 | |||
XM_008761867.1 | XP_008760089.1 | 3 | 158 | |||
GenBank mRNA | BC167754.1 | - | 4 | 413 | 9089038 | |
L19341.1 | - | 3 | 242 |
