siRNA Details
Gene Symbol: Mef2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030860.2 | NP_110487.2 | 3 | 621 | ||
XM_006232771.1 | XP_006232833.1 | 2 | 199 | |||
XM_006232772.1 | XP_006232834.1 | 2 | 199 | |||
XM_006232773.1 | XP_006232835.1 | 2 | 199 | |||
GenBank mRNA | AJ005425.1 | - | 2 | 196 | ||
BC081790.1 | - | 3 | 621 | 7134848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
