siRNA Details
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035255.1 | NP_001030332.1 | 12 | 1947 | ||
XM_006233540.2 | XP_006233602.1 | 13 | 2008 | |||
XM_006233541.2 | XP_006233603.1 | 13 | 1986 | |||
XM_006233542.2 | XP_006233604.1 | 13 | 1984 | |||
XM_006233543.2 | XP_006233605.1 | 14 | 2065 | |||
XM_008761567.1 | XP_008759789.1 | 13 | 2006 | |||
XM_008761568.1 | XP_008759790.1 | 14 | 5056 | |||
XM_008761569.1 | XP_008759791.1 | 14 | 2072 | |||
XM_008761570.1 | XP_008759792.1 | 10 | 2235 | |||
GenBank mRNA | BC099157.1 | - | 12 | 1947 | 7381485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
