siRNA Details
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031052.1 | NP_112314.1 | 6 | 782 | ||
XM_008770779.1 | XP_008769001.1 | 6 | 932 | |||
XM_008770780.1 | XP_008769002.1 | 6 | 932 | |||
GenBank mRNA | M96633.1 | - | 6 | 782 |
