siRNA Details
Gene Symbol: TCAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130026.2 | NP_001123498.2 | 8 | 4460 | ||
XM_006715927.3 | XP_006715990.1 | 7 | 5692 | |||
XM_011516059.2 | XP_011514361.1 | 7 | 4280 | |||
GenBank mRNA | AK090395.1 | - | 6 | 3923 |
Gene Symbol: ATP10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025130.1 | - | 7 | 2039 | ||
AK090832.1 | - | 11 | 2746 |
Gene Symbol: CADM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017459.1 | XP_016872948.1 | 1 | 887 | ||
XM_017017460.1 | XP_016872949.1 | 1 | 887 |
Gene Symbol: ESR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | LC120324.1 | - | 5 | 1315 |
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832127.1 | - | 5 | 2384 |
Gene Symbol: GATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098014.1 | - | 1 | 43 |
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152436.2 | NP_689649.1 | 4 | 1359 | ||
GenBank mRNA | BC029557.1 | - | 4 | 1320 | 5262538 |
Gene Symbol: GPR141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515374.2 | XP_011513676.1 | 7 | 2688 | ||
XM_011515375.2 | XP_011513677.1 | 6 | 2651 | |||
XM_011515383.2 | XP_011513685.1 | 2 | 2017 | |||
XM_011515385.2 | XP_011513687.1 | 3 | 7868 | |||
XM_011515386.2 | XP_011513688.1 | 3 | 2090 | |||
XM_011515388.2 | XP_011513690.1 | 2 | 2236 | |||
XM_011515389.2 | XP_011513691.1 | 2 | 2275 | |||
XM_017012160.1 | XP_016867649.1 | 5 | 2184 | |||
XM_017012161.1 | XP_016867650.1 | 5 | 2438 | |||
XM_017012162.1 | XP_016867651.1 | 6 | 3487 | |||
XM_017012163.1 | XP_016867652.1 | 5 | 3451 | |||
XM_017012164.1 | XP_016867653.1 | 2 | 2025 | |||
XM_017012165.1 | XP_016867654.1 | 3 | 8104 | |||
XM_017012166.1 | XP_016867655.1 | 4 | 3407 | |||
XM_017012167.1 | XP_016867656.1 | 3 | 2117 |
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114396.1 | NP_001107868.1 | 7 | 2771 | ||
NM_002262.3 | NP_002253.1 | 6 | 2834 | |||
NM_007334.2 | NP_031360.1 | 5 | 2741 |
Gene Symbol: LINC00518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027793.1 | 3 | 1793 |
Gene Symbol: MAPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022161.1 | - | 2 | 1558 |
Gene Symbol: OR1J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014623.1 | XP_016870112.1 | 10 | 4236 | ||
GenBank mRNA | AK123016.1 | - | 4 | 3113 |
Gene Symbol: RASAL2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027982.1 | 2 | 1876 | |||
GenBank mRNA | AK124464.1 | - | 2 | 1511 |
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006931.2 | NP_008862.1 | 10 | 2310 | ||
GenBank mRNA | AB209607.1 | - | 9 | 2788 | ||
AK074861.1 | - | 2 | 948 | |||
BC039196.1 | - | 10 | 2185 | 4396508 | ||
M20681.1 | - | 10 | 2290 |
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090510.1 | - | 1 | 1770 |
Gene Symbol: UBE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127227.1 | - | 1 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
