siRNA Details
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031077.1 | NP_112339.1 | 16 | 2845 | ||
GenBank mRNA | U36444.1 | - | 16 | 2845 |

More Information
Other Species Targeted

Gene Ontology Categories:
