siRNA Details
Gene Symbol: SUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005849.1 | NP_001005849.1 | 3 | 342 | ||
NM_006937.3 | NP_008868.3 | 4 | 414 | |||
GenBank mRNA | AK024823.1 | - | 4 | 315 | ||
AK225090.1 | - | 4 | 325 | |||
AK225263.1 | - | 4 | 326 | |||
AK311837.1 | - | 4 | 360 | |||
BC008450.1 | - | 4 | 381 | 4133768 | ||
BC016775.1 | - | 4 | 395 | 4096440 | ||
BC022340.1 | - | 4 | 365 | 4134531 | ||
BC062713.1 | - | 4 | 328 | 30404344 | ||
BC068465.1 | - | 4 | 380 | 5277638 | ||
BC070159.1 | - | 4 | 388 | 6592382 | ||
BC071645.1 | - | 4 | 321 | 3884132 | ||
BC071646.1 | - | 4 | 357 | 4369354 | ||
BC107853.1 | - | 4 | 318 | 5772748 | ||
BF700114.1 | - | 3 | 311 | |||
BG778261.1 | - | 4 | 415 | |||
L76416.1 | - | 4 | 259 | |||
X99585.1 | - | 4 | 335 |
Gene Symbol: LOC105371814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135674.1 | 4 | 650 | |||
GenBank mRNA | CX873301.1 | - | 4 | 653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
