siRNA Details
Gene Symbol: Psen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250405.2 | XP_006250467.1 | 13 | 2105 | ||
XM_006250407.2 | XP_006250469.1 | 12 | 2015 | |||
XM_006250408.2 | XP_006250470.1 | 11 | 1930 | |||
XM_006250410.2 | XP_006250472.1 | 12 | 2695 | |||
XM_008769876.1 | XP_008768098.1 | 12 | 2697 | |||
XM_008769877.1 | XP_008768099.1 | 13 | 2245 | |||
XM_008769878.1 | XP_008768100.1 | 11 | 3323 | |||
GenBank mRNA | X99267.1 | - | 11 | 1977 |
