siRNA Details
Gene Symbol: Cirbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031147.2 | NP_112409.2 | 7 | 774 | ||
XM_006241012.2 | XP_006241074.1 | 6 | 980 | |||
XM_006241013.2 | XP_006241075.2 | 6 | 1091 | |||
XM_006241014.2 | XP_006241076.1 | 7 | 779 | |||
XM_006241015.2 | XP_006241077.2 | 7 | 890 | |||
GenBank mRNA | BC069219.1 | - | 7 | 774 | 6888887 |
