siRNA Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011710.2 | NP_001011710.1 | 7 | 563 | ||
XM_006244643.2 | XP_006244705.1 | 6 | 546 | |||
GenBank mRNA | AB009662.1 | - | 6 | 426 | ||
BC078799.1 | - | 7 | 512 | 7112769 | ||
CK469282.1 | - | 7 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
