siRNA Details
Gene Symbol: UPK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006953.3 | NP_008884.1 | 3 | 407 | ||
GenBank mRNA | AB010116.1 | - | 2 | 284 | ||
AB010637.1 | - | 3 | 380 | |||
AF085808.1 | - | 3 | 412 | |||
BC069544.1 | - | 3 | 412 | 7262218 | ||
BM727658.1 | - | 3 | 386 | |||
CR456608.1 | - | 3 | 382 |
