siRNA Details
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031337.2 | NP_112627.2 | 8 | 1842 | ||
XM_006236676.1 | XP_006236738.1 | 7 | 1503 | |||
XM_006236677.1 | XP_006236739.1 | 8 | 1645 | |||
XM_006236679.2 | XP_006236741.1 | 7 | 1636 | |||
XM_006236680.1 | XP_006236742.1 | 7 | 1519 | |||
XM_006236682.1 | XP_006236744.1 | 6 | 1330 | |||
XM_006236684.2 | XP_006236746.1 | 6 | 1524 | |||
XM_008762996.1 | XP_008761218.1 | 7 | 1665 | |||
XM_008762997.1 | XP_008761219.1 | 6 | 1553 | |||
GenBank mRNA | AB018049.1 | - | 7 | 1508 | ||
BC078798.1 | - | 8 | 1842 | 7106217 |
