siRNA Details
Gene Symbol: FXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000144.4 | NP_000135.2 | 2, 3 | 490 | ||
NM_001161706.1 | NP_001155178.1 | 2, 3 | 490 | |||
NM_181425.2 | NP_852090.1 | 2, 3 | 490 | |||
GenBank mRNA | AK308620.1 | - | 2, 3 | 150 | ||
BC023633.2 | - | 2, 3 | 298 | 4842134 | ||
BC048097.1 | - | 2, 3 | 301 | 5300379 | ||
BF058880.1 | - | 2, 3 | 161 | |||
KU178079.1 | - | 2, 3 | 270 | |||
U43747.1 | - | 2, 3 | 795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
