siRNA Details
Gene Symbol: Chst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053408.1 | NP_445860.1 | 2 | 458 | ||
XM_006256427.2 | XP_006256489.1 | 2 | 572 | |||
XM_006256428.2 | XP_006256490.1 | 3 | 604 | |||
XM_008772929.1 | XP_008771151.1 | 3 | 1109 | |||
GenBank mRNA | AF178689.2 | - | 2 | 458 |
