siRNA Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053420.3 | NP_445872.1 | 6 | 1029 | ||
GenBank mRNA | AF243515.1 | - | 6 | 993 | ||
AY095482.1 | - | 6 | 993 | |||
BC070958.1 | - | 6 | 987 | 7105707 | ||
BC081690.1 | - | 6 | 967 | 7190121 | ||
FQ213448.1 | - | 5 | 790 | |||
FQ215108.1 | - | 6 | 1000 | |||
FQ215127.1 | - | 6 | 1000 | |||
FQ215203.1 | - | 6 | 1020 | |||
FQ216745.1 | - | 6 | 1021 | |||
FQ216768.1 | - | 3 | 606 | |||
FQ217381.1 | - | 1 | 175 | |||
FQ220853.1 | - | 6 | 996 | |||
FQ221849.1 | - | 6 | 1020 |
