siRNA Details
Gene Symbol: Mlycd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053477.1 | NP_445929.1 | 3, 4 | 805 | ||
GenBank mRNA | AF304865.1 | - | 3, 4 | 805 | ||
AJ007704.1 | - | 3, 4 | 805 | |||
BC061845.1 | - | 3, 4 | 808 | 5621015 |
