siRNA Details
Gene Symbol: Hrh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270564.1 | NP_001257493.1 | 3 | 999 | ||
NM_001270565.1 | NP_001257494.1 | 3 | 999 | |||
NM_001270566.1 | NP_001257495.1 | 3 | 999 | |||
NM_001270567.1 | NP_001257496.1 | 3 | 999 | |||
NM_001270568.1 | NP_001257497.1 | 3 | 999 | |||
NM_001270569.1 | NP_001257498.1 | 3 | 999 | |||
NM_001270570.1 | NP_001257499.1 | 3 | 999 | |||
NM_053506.2 | NP_445958.1 | 3 | 999 | |||
XM_008762548.1 | XP_008760770.1 | 2 | 1279 | |||
GenBank mRNA | AB015646.1 | - | 3 | 999 | ||
AF237919.1 | - | 3 | 662 | |||
AY009370.1 | - | 3 | 662 | |||
AY009371.1 | - | 3 | 662 | |||
BC087707.1 | - | 3 | 997 | 7315203 | ||
DQ112342.1 | - | 3 | 662 | |||
DQ112343.1 | - | 3 | 662 | |||
FQ212445.1 | - | 3 | 978 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133849, AK156130, AK140374, AY142145, BC028947, and others.
Set Membership: |
![]() |
Gene Ontology Categories:
