siRNA Details
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053508.2 | NP_445960.1 | 4 | 504 | ||
GenBank mRNA | AJ306427.1 | - | 3 | 433 | ||
BC078773.1 | - | 4 | 504 | 7133616 |

More Information
Other Species Targeted

Gene Ontology Categories:
