siRNA Details
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053528.1 | NP_445980.1 | 3 | 844 | ||
XM_006229410.2 | XP_006229472.1 | 4 | 1185 | |||
XM_008759561.1 | XP_008757783.1 | 4 | 1187 | |||
XM_008759562.1 | XP_008757784.1 | 4 | 1131 | |||
GenBank mRNA | AJ245646.1 | - | 3 | 844 | ||
AJ245647.1 | - | 3 | 844 |
