siRNA Details
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053562.3 | NP_446014.2 | 5 | 501 | ||
GenBank mRNA | AF035673.1 | - | 5 | 503 |

More Information
Other Species Targeted

Gene Ontology Categories:
