siRNA Details
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF063301.1 | - | 1 | 87 | ||
AF205403.1 | - | 2 | 327 | |||
AK314990.1 | - | 2 | 260 | |||
BC032667.2 | - | 1 | 88 | 5541591 | ||
CV571245.1 | - | 2 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
