siRNA Details
Gene Symbol: ATE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001976.2 | NP_001001976.1 | 3, 4 | 355 | ||
NM_001288736.1 | NP_001275665.1 | 3, 4 | 590 | |||
NM_007041.3 | NP_008972.2 | 3, 4 | 355 | |||
NR_110006.1 | 2, 3 | 527 | ||||
XM_005269458.2 | XP_005269515.1 | 3, 4 | 367 | |||
XM_005269459.3 | XP_005269516.1 | 3, 4 | 590 | |||
XM_017015597.1 | XP_016871086.1 | 1, 2 | 69 | |||
XM_017015598.1 | XP_016871087.1 | 3, 4 | 574 | |||
XM_017015599.1 | XP_016871088.1 | 3, 4 | 367 | |||
XM_017015601.1 | XP_016871090.1 | 1, 2 | 69 | |||
XM_017015603.1 | XP_016871092.1 | 1, 2 | 69 | |||
GenBank mRNA | AF079098.1 | - | 3, 4 | 229 | ||
AF079099.1 | - | 3, 4 | 229 | |||
AK124669.1 | - | 2, 3 | 535 | |||
AK299101.1 | - | 3, 4 | 241 | |||
AK313057.1 | - | 3, 4 | 315 | |||
BC022026.1 | - | 3, 4 | 328 | 4825006 | ||
HY046979.1 | - | 3, 4 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
