siRNA Details
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053783.1 | NP_446235.1 | 4 | 524 | ||
GenBank mRNA | AF201901.1 | - | 4 | 524 | ||
BC062039.1 | - | 4 | 466 | 5598500 | ||
U68272.1 | - | 2 | 98 |
