siRNA Details
Gene Symbol: Snap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053810.3 | NP_446262.3 | 4 | 643 | ||
GenBank mRNA | AF035822.1 | - | 4 | 653 | ||
AF260577.1 | - | 4 | 566 | |||
BC091693.1 | - | 4 | 643 | 7134635 |
