siRNA Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053953.1 | NP_446405.1 | 3 | 364 | ||
XM_008766983.1 | XP_008765205.1 | 3 | 749 | |||
XM_008766984.1 | XP_008765206.1 | 3 | 520 | |||
GenBank mRNA | BC091564.1 | - | 3 | 338 | 7445971 | |
FQ228742.1 | - | 3 | 363 | |||
FQ229102.1 | - | 3 | 363 | |||
Z22812.1 | - | 3 | 364 |
