siRNA Details
Gene Symbol: Neu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054010.1 | NP_446462.1 | 2 | 350 | ||
XM_008759664.1 | XP_008757886.1 | 1 | 3773 | |||
XM_008759665.1 | XP_008757887.1 | 2 | 471 | |||
GenBank mRNA | AB026841.1 | - | 2 | 350 | ||
BC100625.1 | - | 2 | 298 | 7935525 |
