siRNA Details
Gene Symbol: Neu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054010.1 | NP_446462.1 | 3 | 955 | ||
XM_008759664.1 | XP_008757886.1 | 2 | 4378 | |||
XM_008759665.1 | XP_008757887.1 | 3 | 1076 | |||
XM_008759666.1 | XP_008757888.1 | 2 | 874 | |||
GenBank mRNA | AB026841.1 | - | 3 | 955 | ||
BC100625.1 | - | 3 | 903 | 7935525 |
