siRNA Details
Gene Symbol: Cflar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057138.2 | NP_476479.1 | 6 | 1186 | ||
GenBank mRNA | AF244366.1 | - | 4 | 976 | ||
CF108550.1 | - | 2 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
